Learn More
Honeywell™ Class 1 Plastic Cartridge A1B1E1 for Organic Vapors and Acid Gas
Product Code. 15876635
Change view
Click to view available options
Protects against organic vapor, acid gas, ammonia and particulates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15876635 Protects against organic vapor, acid gas, ammonia and particulates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15876635 Supplier Honeywell™ Supplier No. N06575089L
  • ONLINE EXCLUSIVE PRICE
    483.00 NOK
    343.54 NOK / 1 pairs
    Save 139.46 NOK
    28% Off
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
The A1B1E1K1P3 cartridge is part of the wide range of cartridges offered to protect against many hazardous gases, vapors and/or particulates in many applications. The range provides high quality and a cost-effective protection.
  • Protects against organic vapor, acid gas, ammonia and particulates
  • Excellent filtration efficiency over extensive period of time
  • Lower profile and lightweight for optimal comfort
  • Low profile does not obstruct the field of vision
  • Threaded connector is easy to attach
TRUSTED_SUSTAINABILITY
Specifications
Cartridge Type Class 1 plastic cartridge
Certifications/Compliance EC Category PPE: 3, QA: ISO 9001/2000, EC Certification: 2259, EC Attestation: 2259, EN 2259
For Use With (Application) Protects against organic vapor, acid gas, ammonia and particulates
Quantity 1 Pair
Weight (Metric) 122.3 g