Learn More
Honeywell™ Class 1 Plastic Cartridge A1B1E1 for Organic Vapors and Acid Gas
Product Code. 15806845
Change view
Click to view available options
Includes threaded connector
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15806845 Includes threaded connector
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15806845 Supplier Honeywell™ Supplier No. N06575008
  • ONLINE EXCLUSIVE PRICE
    103.27 NOK
    88.37 NOK / Pack of 1
    Save 14.90 NOK
    14% Off
Estimated Shipment: 07-12-2026
The P3 cartridge is part of the wide range of high-quality and cost-effective cartridges offered to protect against many hazardous gases, vapors and/or particulates in many applications.
  • Protects against particulates
  • Excellent filtration efficiency over extensive period of time
  • Lower profile and lightweight for optimal comfort
  • Low profile does not obstruct the field of vision
  • Threaded connector is easy to attach
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EC Category PPE: 3, QA: ISO 9001/2000, EC Certification: 0252, EC Attestation: 0252, EN 143, Declaration of conformity INSPEC N06575008
Material ABS - fiberglass paper
For Use With (Application) Agriculture, automotive, aviation, construction, chemical, foundry, general industry, glass, green spaces, industrial cleaning, iron and steel, laboratory, maintenance, medical and pharmaceutical, paper, ship building, textile, utilities, water treatment, welding, wood industries
Includes Threaded connector