Learn More
Honeywell™ N7700 Class 1 Half-Mask Respirator
Description
- 100% Silicone hypoallergenic face piece for extra comfort
- Single unit four-point harness with neck strap is easy to use and its ergonomic design and improved suspension system fits perfectly on top of the head providing a high level of comfort
- Reduced dead air space means less re-breathed exhaled air
- Quick to dismantle and clean, with replaceable parts
- Superior exhalation valve allows more breath to be exhaled so workers feel cooler and more comfortable
- Variable thickness in sealing area conforms to nose bridge for improved comfort and fit
Specifications
Specifications
| Certifications/Compliance | EC Category PPE: 3, EC Certificate: 250, EC Attestation, EN 140 |
| Color | Black |
| Material | Silicone skirt |
| Product Type | Half-mask Respirator |
| Size | Large |
| For Use With (Application) | Suitable for many types of applications |
| Harness Style | 4-point |