missing translation for 'onlineSavingsMsg'
Learn More

Copper 65 Isotope Standard For ICP/MS, SPEX CertiPrep™

Product Code. 11426018 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
125 mL
Packaging:
Triple-leached LDPE bottle
Unit Size:
125mL
This item is not returnable. View return policy

Product Code. 11426018

Brand: SPEX Certiprep ISOTCU65

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Cu 6510μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 7440-50-8, 7732-18-5, 7440-50-8
CAS Max % 2.0, 98.0, 0.0
Physical Form Liquid
Quantity 125 mL
Molecular Formula Cu
MDL Number MFCD00010965
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES [Cu]
IUPAC Name copper
Molecular Weight (g/mol) 63.55
PubChem CID 23978
ChEBI CHEBI:30052
Grade Claritas PPT
Concentration or Composition (by Analyte or Components) 10mg/L
Packaging Triple-leached LDPE bottle
Chemical Name or Material Copper 65 Isotope Standard, For ICP/MS
Show More Show Less
compliance-icons
Product Identifier
  • 10 µg/mL Cu65 Isotope Standard
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : nitric acid

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.