Description
Maximize capacity on the Thermo Scientific™ CO2 Resistant Shaker universal platform with a variety of clamps to fit most vessels.
- Compatible with Thermo Scientific™ CO2Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers.
- Refer to the user manual or brochure for details on the maxinum number of clamps that can fit per platform.
Specifications
Specifications
| Product Type | Starter Kit |
| Clamps | Flask |
| For Use With (Equipment) | CO2 Resistant Shaker |
| Includes | (2) 100mL flask clamps, (4) 250mL flask clamps, (4) 500mL flask clamps, (2) 1L flask clamps, (2) 2L flask clamps |
| No. of Positions | Varies |