Learn More
Thermo Scientific™ Clamps for CO2 Resistant Shaker
Product Code. 15321115 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Flask Clamp
Funnel Clamp
Infusion bottle clamp
Starter Kit
Clamps:
100 mL
1000 mL
150 mL
200 mL
2000 mL
25 mL
250 mL
3000 mL
50 mL
500 mL
5000 mL
Flask
Separatory
Unit Size:
1 unit
Each
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Clamps unitSize
15321115 Flask Clamp 1000 mL Each
15361105 Flask Clamp 50 mL Each
30280804 Flask Clamp 25 mL 1 unit
15371105 Flask Clamp 100 mL Each
15381105 Flask Clamp 150 mL Each
15391105 Flask Clamp 200 mL Each
15301115 Flask Clamp 250 mL Each
15311115 Flask Clamp 500 mL Each
15331115 Flask Clamp 2000 mL Each
15341115 Flask Clamp 3000 mL Each
15351115 Flask Clamp 5000 mL Each
15361115 Infusion bottle clamp 500 mL Each
15371115 Infusion bottle clamp 1000 mL Each
15381115 Funnel Clamp Separatory Each
15321135 Starter Kit Flask Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 15321115 Supplier Thermo Scientific™ Supplier No. 88881111
Only null left
Add to basket
Add to basket
Achieve maximize capacity on the Thermo Scientific™ CO2 Resistant Shaker universal platform with a variety of clamps to fit most vessels.
Maximize capacity on the Thermo Scientific™ CO2 Resistant Shaker universal platform with a variety of clamps to fit most vessels.
  • Compatible with Thermo Scientific™ CO2Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers.
  • Refer to the user manual or brochure for details on the maxinum number of clamps that can fit per platform.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Flask Clamp
Clamps 1000 mL
For Use With (Equipment) CO2 Resistant Shaker
No. of Positions 6