Learn More
Corning™ Clamps for Erlenmeyer Flask
Product Code. p-8007925 Shop All Corning Products
Change view
Click to view available options
Holds:
25 mL Erlenmeyer Flask
50 mL Erlenmeyer Flask
Unit Size:
Each
Product Code. Holds unitSize
11303815 25 mL Erlenmeyer Flask Each
11313815 50 mL Erlenmeyer Flask Each
2 options
Product Code. 11303815
Brand: Corning™ 480110
Only null left
Add to basket
Add to basket
For use with various sizes of Erlenmeyer flasks.
This is a clamp for use with Erlenmeyer flasks, for use with Corning™ LSE™ Orbital Shaker and Corning™ LSE™ Low Speed Orbital Shaker.
TRUSTED_SUSTAINABILITY
Specifications
Holds 25 mL Erlenmeyer Flask
Product Type Clamp