Learn More
DWK Life Sciences™ Clamp for Spherical Joints, with tightening screw, made of stainless steel
Shop All DWK Life Sciences Products
Click to view available options
Size:
S 13
S 19
S 29
S 35
S 40 and S 41
S 51
S 64
Unit Size:
Each
Pack of 10
Pack of 25
Pack of 5
Pack of 50
Description
The ball joint clamp with locking screw enables a very secure connection by additional locking with the locking screw.
- The locking screw enables a very secure connection by means of an additional locking device.
- Excellent dimensional stability compared to conventional wire clamps
Specifications
Specifications
| Quantity | 5 |
| Material | Stainless Steel |
| For Use With (Equipment) | Securing of Spherical Ground Joints |
| For Use With (Application) | Securing of spherical ground joints |
| Size | S 64 |