Learn More
DWK Life Sciences™ Clamp for Spherical Joints, with tightening screw, made of stainless steel
Product Code. 15844678 Shop All DWK Life Sciences Products
Change view
Click to view available options
Size:
S 13
S 19
S 29
S 35
S 40 and S 41
S 51
S 64
Unit Size:
Each
Pack of 10
Pack of 25
Pack of 5
Pack of 50
Product Code. Size unitSize
15844678 S 40 and S 41 Pack of 25
10623982 S 35 Each
15854678 S 51 Pack of 10
10409342 S 29 Each
10141781 S 19 Each
15864678 S 64 Pack of 5
15921571 S 13 Pack of 50
7 options
Product Code. 15844678
Brand: DWK Life Sciences™ 286334605
Only null left
Add to basket
Add to basket
Ball joint clamp with additional locking option.
The ball joint clamp with locking screw enables a very secure connection by additional locking with the locking screw.
  • The locking screw enables a very secure connection by means of an additional locking device.
  • Excellent dimensional stability compared to conventional wire clamps
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25
Material Stainless Steel
For Use With (Equipment) Securing of Spherical Ground Joints
For Use With (Application) Securing of spherical ground joints
Size S 40 and S 41