Learn More
OHAUS™ Clamp Connector
Shop All Ohaus Products
Click to view available options
Connector Types:
Bar
Channel
End-To-End
Hook
Lattice
Multi-Rod
Rod End
S
Material:
Aluminum
Nickel-plated
Stainless Steel
Size:
0 to 13 mm (Grip Range)
102 mm (L)
152 mm (L)
203 mm (L)
40 x 33 mm (H x L)
51 mm (L)
missing translation for 'unitSize'
Each
Description
- Hold glassware safely
- Corrosion-resistant
- Providing convenience with exceptional stability and durability
Specifications
Specifications
| Connector Types | Rod End |
| Type | Connector |
| Material | Nickel-plated |
| Includes | Fixed 90° Connection Angle |
| Description | Clamp, Connector, Rod End, CLC-RDENDZ |
| Size | 0 to 13 mm (Grip Range) |
| Size (Metric) Grip | 0 to 13 mm |