Learn More
OHAUS™ Clamp Connector
Product Code. 15752309 Shop All Ohaus Products
Click to view available options
Connector Types:
Bar
Channel
End-To-End
Hook
Lattice
Multi-Rod
Rod End
S
Material:
Aluminum
Nickel-plated
Stainless Steel
Size:
0 to 13 mm (Grip Range)
102 mm (L)
152 mm (L)
203 mm (L)
40 x 33 mm (H x L)
51 mm (L)
Unit Size:
Each
Product Code. 15752309
Brand: OHAUS™ 30392261
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
OHAUS™ Clamp Connector provides secure support for holding glassware at elevated positions.
  • Hold glassware safely
  • Corrosion-resistant
  • Providing convenience with exceptional stability and durability
TRUSTED_SUSTAINABILITY
Specifications
Connector Types Channel
Type Connector
Material Nickel-plated
Description Clamp, Connector, Channel, CLC-CHANLZ
Height (Metric) 40.000 mm
Height (English) 1.58 in.
Size 40 x 33 mm (H x L)
Width (English) 1.30 in.
Width (Metric) 33.00 mm