Description
- Hold glassware safely
- Corrosion-resistant
- Providing convenience with exceptional stability and durability
Specifications
Specifications
| Connector Types | S |
| Type | Connector |
| Material | Nickel-plated |
| Includes | Fixed 90° Connection Angle |
| Description | Clamp, Connector, S, CLC-SCONNZ |
| Size | 0 to 13 mm (Grip Range) |
| Size (Metric) Grip | 0 to 13 mm |