missing translation for 'onlineSavingsMsg'
Learn More

CL-82198, MedChemExpress

Product Code. p-200097240
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16482556 25 mg 25mg
16425707 5 mg 5mg
16419536 50 mg 50mg
16414286 100 mg 100mg
16402968 10 mg 10mg
5 options
This item is not returnable. View return policy

Product Code. 16482556

Brand: Medchem Express HY100359

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CL-82198 is a selective inhibitor of MMP-13. CL-82198 binds to the entire S1’ pocket of MMP-13, which is the basis for its selectivity towards MMP-13 and the lack of inhibitory activities against other MMPs. CL-82198 is a pharmacologic treatment for preventing osteoarthritis (OA) progression.

CL-82198 is a selective inhibitor of MMP-13. CL-82198 binds to the entire S1’ pocket of MMP-13, which is the basis for its selectivity towards MMP-13 and the lack of inhibitory activities against other MMPs. CL-82198 is a pharmacologic treatment for preventing osteoarthritis (OA) progression.

Specifications

Chemical Name or Material CL-82198
Physical Form Solid
CAS 307002-71-7
Color Yellow
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C17H22N2O3
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 100 mg/mL (330.72 mM; Need ultrasonic)
SMILES O=C(C1=CC2=CC=CC=C2O1)NCCCCN3CCOCC3
Molecular Weight (g/mol) 302.37
Formula Weight 302.37
Percent Purity 99.78%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.