missing translation for 'onlineSavingsMsg'
Learn More

CL-387785, MedChemExpress

Product Code. 16400086
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16400086 2 mg 2mg
16486537 5 mg 5mg
16462866 25 mg 25mg
16424866 100 mg 100mg
16423818 10 mg 10mg
16400197 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16400086 Supplier Medchem Express Supplier No. HY10325
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress CL-387785(EKI785; WAY-EKI 785) is an irreversible inhibitor of EGFR with IC50 of 370 pM.

CL-387785(EKI785

Specifications

Chemical Name or Material CL-387785
CAS 194423-06-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H13BrN4O
Quantity 2 mg
Synonym EKI-785 WAY-EKI 785
Solubility Information DMSO : 13.67 mg/mL (35.86 mM; Need ultrasonic and warming)
SMILES O=C(NC1=CC2=C(NC3=CC=CC(Br)=C3)N=CN=C2C=C1)C#CC
Molecular Weight (g/mol) 381.23
Formula Weight 381.23
Percent Purity 98.1%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.