missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CKB Mouse anti-Human, Clone: 2B7C3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66764-1-IG-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.Specifications
| CKB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P12277 | |
| CKB | |
| CKB Fusion Protein Ag7933 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 2B7C3 | |
| Unconjugated | |
| CKB | |
| B CK, CKB, CKBB, Creatine kinase B chain, Creatine kinase B type, creatine kinase, brain | |
| Mouse | |
| Protein A | |
| RUO | |
| 1152 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction