missing translation for 'onlineSavingsMsg'
Learn More

Citrate synthase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16885644 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16885644 150 μL 150µL
16895644 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16885644

Brand: Proteintech 161311AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is specific to CS.

The protein encoded by this gene is a Krebs tricarboxylic acid cycle enzyme that catalyzes the synthesis of citrate from oxaloacetate and acetyl coenzyme A. The enzyme is found in nearly all cells capable of oxidative metablism. This protein is nuclear encoded and transported into the mitochondrial matrix, where the mature form is found.

Specifications

Antigen Citrate synthase
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CS
Gene Accession No. O75390, Q8VHF5, Q9CZU6
Gene Alias Citrate synthase, CS
Gene Symbols CS
Host Species Rabbit
Immunogen Citrate synthase Fusion Protein Ag9117
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12974, 1431, 170587
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.