Learn More
Citrate Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™
Shop All Fisher Chemical Products
Specifications
Specifications
| Chemical Name or Material | Citrate |
| Packaging | Bottle |
| Physical Form | Liquid |
| Molecular Formula | C6H5O73- |
| Concentration | 1 g/L |
| Type | Single-component Standard Solution |
| Solvent or Matrix | Water |
| Quantity | 500 mL |