Learn More
Citraconic Anhydride, Sigma-Aldrich™
Specifications
Specifications
| CAS | 616-02-4 |
| Melting Point | 7°C to 8°C |
| Density | 1.243 g/cm3 at 20°C |
| Boiling Point | 213°C to 214°C/1013 hPa |
| MDL Number | MFCD00005522 |
| Quantity | 100 mL |
| Synonym | Citraconic anhydride, Methylmaleic anhydride, Citraconic anhydride |
| Molecular Weight (g/mol) | 112.08 |
| Grade | Research |