missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ cis VH 298

Negative control for VH 298
Brand: Tocris Bioscience™ 6157/10
This item is not returnable.
View return policy
Description
cis VH 298 is a negative control for VH 298 (Cat.No. 6156). Exhibits no detectable binding of VHL.Specifications
| PROTAC Controls | |
| C27H33N5O4S | |
| Store at -20°C | |
| 10 mg |
| HIF | |
| 523.65g/mol | |
| HIF Activators |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only