Learn More
Alfa Aesar™ cis-Dichlorobis(pyridine)platinum(II), 99%, Thermo Scientific™
Product Code. 11419917
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419917 0.25 g 0.25g
11409917 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11419917 Supplier Alfa Aesar™ Supplier No. B22865.01
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14872-21-0
Molecular Formula C10H10Cl2N2Pt
Molecular Weight (g/mol) 424.19
MDL Number MFCD00058858
InChI Key FCJXCLMZOWAKNQ-UHFFFAOYSA-L
Synonym dichloroplatinum; pyridine, platinum cis-dichlorobispyridine, platinum ii chloride; bis pyridine, dichlorobis pyridine platinum ii, cis-dichlorobis pyridine platinum ii, platinum, dichloro-, compd. with pyridine 1:2
PubChem CID 498600
IUPAC Name dichloroplatinum;pyridine
SMILES Cl[Pt]Cl.C1=CC=NC=C1.C1=CC=NC=C1
Specifications
Melting Point 282°C to 288°C
Quantity 0.25 g
Formula Weight 424.19
Percent Purity 99%
Chemical Name or Material cis-Dichlorobis(pyridine)platinum(II)