Learn More
Labconco™ Circulation Tubing Kit
Click to view available options
Diameter (English) Inside:
0.75 in.
Unit Size:
Each
Description
- Two 2cm (0.75 in.) I. D. flexible neoprene tubes
- Brass fittings 2cm (0.75 in.) O.D. on each end of tubing
- Includes two sections of tubing with fitting
Specifications
Specifications
| Product Type | Tubing Kit |
| Diameter (English) Inside | 0.75 in. |