Learn More
Heinz Herenz Circular Cover Glasses
Description
- Round cover glasses
Specifications
Specifications
| Product Type | Coverslip |
| Shape | Circle |
| Material | Borosilicate Glass |
| Width (Metric) | 10 mm |
| Thickness (Metric) | 1 mm |
| Sterility | Non-sterile |
| Disposable | Single-use |