Learn More
CIRBP Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10209-2-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16846443
  • 3828.53 NOK / 150µL
Estimated Shipment: 13-10-2023
Description
The CIRBP antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CIRBP. This antibody recognizes human, mouse, rat antigen. The CIRBP antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Specifications
Specifications
CIRBP
Polyclonal
Unconjugated
150 μL
1153
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Co-immunoprecipitation (Co-IP)
293 μg/mL
Rabbit
Primary
Human, Rat, Mouse