missing translation for 'onlineSavingsMsg'
Learn More

CIRBP Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Product Code. 16856443 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16856443 20 μL 20µL
16846443 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16856443

Brand: Proteintech 102092AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The CIRBP antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CIRBP. This antibody recognizes human, mouse, rat antigen. The CIRBP antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.

Specifications

Antigen CIRBP
Applications Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Co-immunoprecipitation (Co-IP)
Classification Polyclonal
Concentration 293 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 1153
Target Species Human, Rat, Mouse
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.