Learn More
CIRBP Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10209-2-AP-150UL
Description
The CIRBP antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CIRBP. This antibody recognizes human, mouse, rat antigen. The CIRBP antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Specifications
| CIRBP | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 1153 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Co-immunoprecipitation (Co-IP) | |
| 293 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |