missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CINP Polyclonal antibody specifically detects CINP in Human,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | CINP |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 405 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CDK2-interacting protein, cyclin-dependent kinase 2 interacting protein, cyclin-dependent kinase 2-interacting protein, MGC849 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-212 of human CINP (NP_116019.1).,, Sequence:, MEAKTLGTVTPRKPVLSVSARKIKDNAADWHNLILKWETLNDAGFTTANNIANLKISLLNKDKIELDSSSPASKENEEKVCLEYNEELEKLCEELQATLDGLTKIQVKMEKLSSTTKGICELENYHYGEESKRPPLFHTWPTTHFYEVSHKLLEMYRKELLLKRTVAKELAHTGDPDLTLSYLSMWLHQPYVESDSRLHLESMLLETGHRAL |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?