missing translation for 'onlineSavingsMsg'
Learn More

CIN85 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12132-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16849663

  • 3828.53 NOK / 150µL
Estimated Shipment: 12-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes an adapter protein that contains three N-terminal Src homology domains, a proline rich region and a C-terminal coiled-coil domain. The encoded protein facilitates protein-protein interactions and has been implicated in numerous cellular processes including apoptosis, cytoskeletal rearrangement, cell adhesion and in the regulation of clathrin-dependent endocytosis. Alternate splicing results in multiple transcript variants.
Specifications

Specifications

CIN85
Polyclonal
Unconjugated
SH3KBP1
CD2 binding protein 3, CD2BP3, CIN85, GIG10, HSB 1, MIG18, SH3KBP1
Rabbit
Antigen Affinity Chromatography
RUO
30011, 58194, 84357
-20°C
Liquid
Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8R550, Q925Q9, Q96B97
Sh3kbp1
CIN85 Fusion Protein Ag2777
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers