missing translation for 'onlineSavingsMsg'
Learn More

CIN85 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16859663 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16859663 20 μL 20µL
16849663 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16859663

Brand: Proteintech 121321AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes an adapter protein that contains three N-terminal Src homology domains, a proline rich region and a C-terminal coiled-coil domain. The encoded protein facilitates protein-protein interactions and has been implicated in numerous cellular processes including apoptosis, cytoskeletal rearrangement, cell adhesion and in the regulation of clathrin-dependent endocytosis. Alternate splicing results in multiple transcript variants.

Specifications

Antigen CIN85
Applications Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SH3KBP1
Gene Accession No. Q8R550, Q925Q9, Q96B97
Gene Alias CD2 binding protein 3, CD2BP3, CIN85, GIG10, HSB 1, MIG18, SH3KBP1
Gene Symbols Sh3kbp1
Host Species Rabbit
Immunogen CIN85 Fusion Protein Ag2777
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30011, 58194, 84357
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.