Learn More
Thermo Scientific™ Cimarec™ Extension Cables
Product Code. 10185411 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
4-pin Extension Cable
6-pin Extension Cable
8-pin Extension Cable
Unit Size:
Each
Product Code. Type unitSize
10185411 4-pin Extension Cable Each
10013881 6-pin Extension Cable Each
10013431 8-pin Extension Cable Each
3 options
Product Code. 10185411
Brand: Thermo Scientific™ 50088016
Only null left
Add to basket
Add to basket
Use Thermo Scientific™ Cimarec™ Extension Cables with Cimarec Magnetic Stirrers.
Use Thermo Scientific™ Cimarec™ Extension Cables with Cimarec Magnetic Stirrers.
  • Length: 3m (9.8′)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Telemodul 20 C, 40 C and 10 M
Length (Metric) 3 m
Type 4-pin Extension Cable