Learn More
Thermo Scientific™ Cimarec™ 40M Controllers
Maintain constant speeds, even during viscosity changes, with our 40M controllers.
Brand:  Thermo Scientific™ 50118911
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 16615495
  • 21644.77 NOK / Each
Estimated Shipment: 26-06-2024
Description
Maintain constant speeds, even during viscosity changes, with Thermo Scientific™ Cimarec™ 40M Controllers. This product is required for use with the Thermo Scientific™ Cimarec™ Mobil Direct.
Features:
  • Adjustable speeds from 100 to 1000rpm
  • High visibility digital display
  • Adjustable start-up times
  • Thermal and electrical overload protection, with fault alert
Telemodul 40 M
  • Powerful controller equips Mobil 200 and 600 stirrers for the most demanding mixing tasks
Telemodul 80 M
  • Offers all the functionality of the Telemodul 40 M, with power output boosted to 80W for more demanding stirring tasks
Specifications
Specifications
Telemodul 80 M Control Units (80w)
Stirrer Controller
230 V
100 to 1000 rpm
230 V UK
100 RPM to 1000 RPM
Special Offers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.