Learn More
CIM 0216, TRC
Product Code. 17910745
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17910745 5 mg 5mg
17951765 10 mg 10mg
17921345 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17910745 Supplier Toronto Research Chemicals Supplier No. TRCC4415505MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1031496-06-6
Molecular Formula C21 H21 N3 O2
Molecular Weight (g/mol) 347.41
Synonym 3,4-Dihydro-N-(5-methyl-3-isoxazolyl)-α-phenyl-1(2H)-quinolineacetamide, CIM 0216
IUPAC Name 2-(3,4-dihydro-2H-quinolin-1-yl)-N-(5-methylisoxazol-3-yl)-2-phenyl-acetamide
SMILES Cc1onc(NC(=O)C(N2CCCc3ccccc23)c4ccccc4)c1
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C21H21N3O2/c1-15-14-19(23-26-15)22-21(25)20(17-9-3-2-4-10-17)24-13-7-11-16-8-5-6-12-18(16)24/h2-6,8-10,12,14,20H,7,11,13H2,1H3,(H,22,23,25)
Formula Weight 347.1634
Chemical Name or Material 3,4-Dihydro-N-(5-methyl-3-isoxazolyl)-alpha-phenyl-1(2H)-quinolineacetamide