missing translation for 'onlineSavingsMsg'
Learn More

Cilofexor, MedChemExpress

Product Code. p-200096101
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16483016 25 mg 25mg
16426997 5 mg 5mg
16420607 50 mg 50mg
16415206 100 mg 100mg
16404278 10 mg 10mg
5 options
This item is not returnable. View return policy

Product Code. 16483016

Brand: Medchem Express HY109083

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cilofexor (GS-9674) is a potent, selective and orally active nonsteroidal FXR agonist with an EC50 of 43 nM. Cilofexor has anti-inflammatory and antifibrotic effects. Cilofexor has the potential for primary sclerosing cholangitis (PSC) and nonalcoholic steatohepatitis (NASH) research.

Cilofexor (GS-9674) is a potent, selective and orally active nonsteroidal FXR agonist with an EC50 of 43 nM. Cilofexor has anti-inflammatory and antifibrotic effects. Cilofexor has the potential for primary sclerosing cholangitis (PSC) and nonalcoholic steatohepatitis (NASH) research.

Specifications

Chemical Name or Material Cilofexor
CAS 1418274-28-8
Color White
Purity Grade Notes Research
Molecular Formula C28H22Cl3N3O5
Quantity 25 mg
Synonym GS-9674
Solubility Information DMSO : 50 mg/mL (85.20 mM; Need ultrasonic)
SMILES ClC1=C(C2(CN(C3=CC(C(O)=O)=CC=N3)C2)O)C=CC(OCC4=C(C5CC5)ON=C4C(C(Cl)=CC=C6)=C6Cl)=C1
Molecular Weight (g/mol) 586.85
Formula Weight 586.85
Percent Purity 98.25%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.