Learn More
Novus Biologicals™ CIITA Antibody Blocking Peptide
Product Code. 18295244 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
Product Code. Quantity unitSize
18295244 0.05 mg 0.05mg
1 options
Product Code. 18295244
Brand: Novus Biologicals™ NBP259072PEP
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
16 amino acids near the amino terminus of human CIITA. Source: Synthetic (Accession #: P33076) The CIITA Blocking Peptide has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY
Specifications
Gene ID (Entrez) 4261
Protein Family CIITA
Common Name CIITA Blocking Peptide
Concentration 0.2 mg/mL
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin.
For Use With (Application) Antibody Competition
Gene Alias C2TA, C2TAMHC class II transactivator type III, CIITAIV, class II, major histocompatibility complex, transactivator, MHC class II transactivator, MHC2TANLR family, acid domain containing, NLRA, nucleotide-binding oligomerization domain, leucine rich repea
Gene Symbol CIITA
Label Type Unlabeled
Product Type Recombinant Protein
Quantity 0.05 mg
Regulatory Status RUO
Source Synthetic
Show More Show Less
For Research Use Only.