missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIDEA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
1975.00 NOK - 2985.00 NOK
Specifications
| Antigen | CIDEA |
|---|---|
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18618025
|
Novus Biologicals
NBP2-98783-100ul |
100 μg |
2985.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18626125
|
Novus Biologicals
NBP2-98783-50ul |
50 μg |
1975.00 NOK
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CIDEA Polyclonal antibody specifically detects CIDEA in Human samples. It is validated for Western BlotSpecifications
| CIDEA | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS (pH 7.0) | |
| 1149 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| cell death activator CIDE-A, cell death-inducing DFFA-like effector aCIDE-A | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CIDEA. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title