Learn More
CIB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11823-1-AP-150UL
 View more versions of this product
Product Code. 16849153
  • 4836.10 NOK / 150µL
Description
The protein encoded by this gene is a member of the calcium-binding protein family. The specific function of this protein has not yet been determined; however this protein is known to interact with DNA-dependent protein kinase and may play a role in kinase-phosphatase regulation of DNA end joining. This protein also interacts with integrin alpha(IIb)beta(3), which may implicate this protein as a regulatory molecule for alpha(IIb)beta(3).
Specifications
Specifications
CIB1
Polyclonal
Unconjugated
CIB1
Calmyrin, CIB, CIB1, CIBP, DNA PKcs interacting protein, Kinase interacting protein, KIP, KIP1, PRKDCIP, SIP2 28, SNK interacting protein 2 28
Rabbit
Antigen Affinity Chromatography
RUO
10519, 23991, 81823
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q99828, Q9R010, Q9Z0F4
CIB1
CIB1 Fusion Protein Ag2391
150 μL
Primary
Human, Mouse, Rat