Learn More
CIB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11823-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the calcium-binding protein family. The specific function of this protein has not yet been determined; however this protein is known to interact with DNA-dependent protein kinase and may play a role in kinase-phosphatase regulation of DNA end joining. This protein also interacts with integrin alpha(IIb)beta(3), which may implicate this protein as a regulatory molecule for alpha(IIb)beta(3).Specifications
| CIB1 | |
| Polyclonal | |
| Unconjugated | |
| CIB1 | |
| Calmyrin, CIB, CIB1, CIBP, DNA PKcs interacting protein, Kinase interacting protein, KIP, KIP1, PRKDCIP, SIP2 28, SNK interacting protein 2 28 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10519, 23991, 81823 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q99828, Q9R010, Q9Z0F4 | |
| CIB1 | |
| CIB1 Fusion Protein Ag2391 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |