Learn More
cIAP2 Rabbit anti-Human, Polyclonal, Proteintech
Product Code. 16842785 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16842785 20 μL 20µL
16832785 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16842785 Supplier Proteintech Supplier No. 243041AP20UL
Rabbit Polyclonal Antibody
The protein encoded by this gene is a member of a family of proteins that inhibits apoptosis by binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2, probably by interfering with activation of ICE-like proteases. The encoded protein inhibits apoptosis induced by serum deprivation but does not affect apoptosis resulting from exposure to menadione, a potent inducer of free radicals. The amino acid sequence predicts three baculovirus IAP repeat domains and a ring finger domain. Transcript variants encoding the same isoform have been identified.
Specifications
Antigen cIAP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene BIRC3
Gene Accession No. Q13489
Gene Alias AIP1, API2, Apoptosis inhibitor 2, BIRC3, C IAP2, CIAP2, HAIP1, hIAP 1, HIAP1, IAP 1, IAP homolog C, IAP1, MALT2, MIHC, RING finger protein 49, RNF49
Gene Symbols BIRC3
Host Species Rabbit
Immunogen cIAP2 Fusion Protein Ag20143
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 330
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG