missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cIAP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10022-1-AP-20UL
This item is not returnable.
View return policy
Description
This BIRC2 antibody (10022-1-AP) can bind both full length (70 kDa) and cleaved form (60 kDa) of the protein.
The protein encoded by this gene is a member of a family of proteins that inhibits apoptosis by binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2, probably by interfering with activation of ICE-like proteases. This encoded protein inhibits apoptosis induced by serum deprivation and menadione, a potent inducer of free radicals.Specifications
| cIAP1 | |
| Polyclonal | |
| Unconjugated | |
| BIRC2 | |
| API1, BIRC2, C IAP1, cIAP1, Hiap 2, HIAP2, IAP 2, IAP homolog B, IAP2, MIHB, RING finger protein 48, RNF48 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11797, 329 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13490, Q62210 | |
| BIRC2 | |
| cIAP1 Fusion Protein Ag15203 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction