missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIAO1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10295-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
10295-1-AP detected 48-55 kDa band because of phosphorylation. 10295-1-AP does well in IHC and IF detection.
WDR39 is a member of the WD40 family of proteins. WDR39 specifically interacts with WT1 both in vitro and in vivo. This interaction results in a decrease in transcriptional activation mediated by WT1. WDR39 does not inhibit binding of WT1 to its consensus nucleotide sequence and does not affect the repression activity of WT1. Thus, WDR39 appears to specifically modulate the transactivation activity of WT1 and may function to regulate the physiological functions of WT1 in cell growth and differentiation.Specifications
CIAO1 | |
Polyclonal | |
Unconjugated | |
CIAO1 | |
CIA1, CIAO1, WDR39 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9391 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O76071 | |
CIAO1 | |
CIAO1 Fusion Protein Ag0349 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |