Learn More
CHZ868, MedChemExpress
Product Code. 16483667
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16483667
Brand: Medchem Express HY18960
MedChemExpress CHZ868 is a type II JAK2 inhibitor with an IC50 of 0.17 μM in EPOR JAK2 WT Ba/F3 cell.
CHZ868 is a type II JAK2 inhibitor with an IC50 of 0.17 μM in EPOR JAK2 WT Ba/F3 cell.
Specifications
Chemical Name or Material CHZ868
CAS 1895895-38-1
Color Gray
Purity Grade Notes Research
Molecular Formula C22H19F2N5O2
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (236.17 mM; Need ultrasonic)
SMILES CC(NC1=CC(OC2=CC=C(N(C)C(NC3=CC=C(F)C=C3F)=N4)C4=C2C)=CC=N1)=O
Molecular Weight (g/mol) 423.42
Formula Weight 423.42
Percent Purity 98.67%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.