missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHST15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | CHST15 |
|---|---|
| Concentration | 0.23 mg/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16872874
|
Proteintech
14298-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16862874
|
Proteintech
14298-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GALNAC4S-6ST is a sulfotransferase that transfers sulfate from 3′-phosphoadenosine 5′-phosphosulfate (PAPS) to the C-6 hydroxyl group of the GalNAc 4-sulfate residue of chondroitin sulfate A and forms chondroitin sulfate E containing GlcA-GalNAc(4, 6-SO(4)) repeating units. It also transfers sulfate to a unique non-reducing terminal sequence, GalNAc(4SO4)-GlcA(2SO4)-GalNAc(6SO4), to yield a highly sulfated structure similar to the structure found in thrombomodulin chondroitin sulfate. GALNAC4S-6ST may also act as a B-cell receptor involved in BCR ligation-mediated early activation that mediate regulatory signals key to B-cell development and/or regulation of B-cell-specific RAG expression; however such results are unclear in vivo.Specifications
| CHST15 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| Q7LFX5, Q8CHI9, Q91XQ5 | |
| 286974, 51363, 77590 | |
| CHST15 Fusion Protein Ag5679 | |
| Primary | |
| -20°C | |
| CHST15 |
| 0.23 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| BRAG, CHST15, GALNAC4S 6ST, GALNAC4S6ST, hBRAG, KIAA0598, RP11 47G11.1 | |
| Chst15 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title