Learn More
Thermo Scientific™ Chromogenic Cronobacter Isolation Agar
Product Code. 15802447
Click to view available options
Quantity:
10 Plates
Unit Size:
Pack of 10
Product Code. 15802447
Brand: Thermo Scientific™ PO1242A
Only null left
Add to basket
Add to basket
Ensure accurate isolation of Cronobacter species with this agar, providing clear differentiation and reliable results.
Ensure accurate isolation of Cronobacter species with Chromogenic Cronobacter Isolation Agar, providing clear differentiation and reliable results.
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
Content And Storage 2°C to 12°C
Format 90 mm
Manufacturing Origin United Kingdom
Product Type Agar Plate
Fill Volume 17 mL
Quantity 10 Plates