missing translation for 'onlineSavingsMsg'
Learn More

Chromium Standard: Cr @ 10000 μg/mL in 5% HNO3

Product Code. 30324153
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
Product Code. Quantity unitSize
30324153 500 mL 500mL
30310328 250 mL 250mL
30314798 100 mL 100mL
3 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 30324153

Brand: VHG Labs VHGTCRN500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chromium
CAS 7440-47-3
Analyte or Component Names Chromium
Shipping Condition Room Temperature
Molecular Formula Cr
Concentration 10000 μg/mL
Certifications/Compliance ISO 17034
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Synonym Cr
InChI Formula InChI=1 S/Cr
Solution Type Single Solution
SMILES [Cr]
Element Cr
IUPAC Name Chromium
Solvent or Matrix Dilute HNO3
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 52
Quantity 500 mL
Formula Weight 51.941
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.