Learn More
Chromium for AAS 1g/L HCl matrix, Fisher chemical™
BIOPHARMA
Product Code. 12903145 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
1000 mL
Packaging:
Bottle
Unit Size:
100mL
500mL
Product Code. 12903145
Brand: Fisher Chemical B 013 500
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Chemical Identifiers
CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O
Specifications
Packaging Bottle
Physical Form Liquid
Concentration 1 g/L ±0.002 g/L
Type Flame Single-element Standard Solution
Solvent or Matrix 2% HCl
Quantity 1000 mL
Chemical Name or Material Chromium