Description
Includes:
- Data Client for data processing
- Compliance tools, enabling user management, object audit trails, versioning and electronic signatures
- Instrument Operation (both locally and remotely)
- Virtual Column Complete, enabling modeling of both isocratic and linear gradient separations
- Report Designer Pro for creation and/or charting of custom Excel-style formulas.
Specifications
Specifications
| Type | Chromatography Software |
| For Use With (Equipment) | Chromeleon CDS |
| Certifications/Compliance | 21 CFR |
| Product Line | Chromeleon 7 |
| Operating System | Windows 11 |
| Quantity | 1 Ea. |