Learn More
Bochem™ Chrome Steel PVC Coated 3-Fingers Clamps
Product Code. 10460923
Change view
Click to view available options
Width (Metric):
0 to 35 mm
0 to 60 mm
Unit Size:
Each
Product Code. Width (Metric) unitSize
10460923 0 to 60 mm Each
10556951 0 to 35 mm Each
2 options
Product Code. 10460923
Brand: Bochem™ 5627
Only null left
Add to basket
Add to basket
Includes shaft. Bochem™ Chrome Steel PVC Coated 3-Fingers Clamps span width independently adjustable by 2 regulating screws.
  • Finger PVC coated
  • Span width independently adjustable by 2 regulating screws
  • With shaft
Specifications
Quantity 1 Ea.
Type 3-Prong Clamp
Material Chrome Steel
Length (Metric) 150 mm
Width (Metric) 0 to 60 mm
Diameter (Metric) Inside 10 mm