Learn More
Macherey-Nagel™ Chrombond™ RS 4 C18 ec Polypropylene Flash Column
Product Code. 12877501 Shop All Macherey-Nagel Products
Change view
Click to view available options
I.D.: 12.4mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12877501 I.D.: 12.4mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12877501 Supplier Macherey-Nagel™ Supplier No. 732810
In Stock
Specifications
Length (Metric) 98 mm
Material Polypropylene
Type Flash Column
Column Type Flash Column
For Use With (Equipment) Directly Used in The Teledyne Isco Companion, Rf