Learn More
Macherey-Nagel™ Chrombond™ RS 330 SiOH Polypropylene Flash Column
Shop All Macherey-Nagel Products
Click to view available options
:
I.D.: 60mm
Description
- For convenient operation and reliable upscaling the complete program of ready-to-use Flash cartridges Female Luer lock inlet and male Luer outlet
- From 4 g to 1600 g adsorbent from one of the leading companies in silica and TLC business
- Increases flexibility with a program of different phases and modifications
- Saves time and money
- Increases analytical safety
Specifications
Specifications
| Length (Metric) | 270 mm |
| Material | Polypropylene |
| Type | Flash Column |
| Column Type | Flash Column |
| For Use With (Equipment) | Directly Used in The Teledyne Isco Companion, Rf |