Learn More
Macherey-Nagel™ Chrombond™ RS 25 NH2 Polypropylene Flash Column
Product Code. 12707760 Shop All Macherey-Nagel Products
Change view
Click to view available options
:
I.D.: 21.2mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12707760 I.D.: 21.2mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12707760 Supplier Macherey-Nagel™ Supplier No. 732822
Only null left
Specifications
Length (Metric) 165 mm
Material Polypropylene
Type Flash Column
Column Type Flash Column
For Use With (Equipment) Directly Used in The Teledyne Isco Companion, Rf