Learn More
Macherey-Nagel™ Chrombond™ RS 25 C18 ec Polypropylene Flash Column
Product Code. 12728332 Shop All Macherey-Nagel Products
Change view
Click to view available options
Length (Metric):
165 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12728332 165 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12728332 Supplier Macherey-Nagel™ Supplier No. 732812
  • ONLINE EXCLUSIVE PRICE
    1720.00 NOK
    1623.90 NOK / Pack of 1
    Save 96.10 NOK
    5% Off
View all promotions
In Stock
Specifications
Length (Metric) 165 mm
Material Polypropylene
Type Flash Column
Column Type Flash Column
For Use With (Equipment) Directly Used in The Teledyne Isco Companion, Rf