Learn More
Macherey-Nagel™ Chrombond™ RS 200 C18 ec Polypropylene Flash Column
Product Code. 10742247 Shop All Macherey-Nagel Products
Change view
Click to view available options
I.D.: 60mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10742247 I.D.: 60mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10742247 Supplier Macherey-Nagel™ Supplier No. 732816
In Stock
Specifications
Length (Metric) 200 mm
Material Polypropylene
Type Flash Column
Column Type Flash Column
For Use With (Equipment) Directly Used in The Teledyne Isco Companion, Rf